Order Entry
Export
ContactUsLinkComponent
2,3-Difluorobenzoic acid, Sigma-Aldrich®
Irritant
2,3-Difluorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL263516-10G
Supplier:  Merck
CAS Number:  
Irritant
2,3-Difluorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL263516-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :158.1 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:4519-39-5
Formula:C₇H₄F₂O₂
MDL Number:MFCD00010267