2,6-Difluorobenzamide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:18063-03-1
:
Formula:C₇H₅F₂NO
MDL Number:MFCD00007972
:157.12 g/mol
Boiling Pt:284 °C (1013 hPa)
Melting Pt:144…145.5 °C
:M-Clarity™ quality level = MQ100
EINECS:241-972-8