3,5-Difluorobenzonitrile, Sigma-Aldrich®
Catalog # SIAL290203-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:139.1 g/mol
Boiling Pt:159…160 °C (1013 hPa)
UN:3439
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:264-752-3
CAS Number:64248-63-1
Formula:C₇H₃F₂N
MDL Number:MFCD00010311