3,4-Difluorobenzonitrile, Sigma-Aldrich®
SIAL264334-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:139.1 g/mol
:189 °C (1013 hPa)
:51 °C
:6.1,III
:M-Clarity™ quality level = MQ200
:264-751-8
:3439
:64248-62-0
:C₇H₃F₂N
:MFCD00011666