Order Entry
Export
ContactUsLinkComponent
3,5-Difluorobenzaldehyde, Sigma-Aldrich®
Irritant
3,5-Difluorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL290173-5G
Supplier:  Merck
CAS Number:  
Irritant
3,5-Difluorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL290173-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :142.1 g/mol
Boiling Pt:61…63 °C (43 mmHg)
Storage Temperature:Ambient
ADR:3,III
 :M-Clarity™ quality level = MQ100
UN:1989
 :GHS07
CAS Number:32085-88-4
Formula:C₇H₄F₂O
MDL Number:MFCD00010329