Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluorophenylboronic acid
3,5-Difluorophenylboronic acid, Sigma-Aldrich®
3,5-Difluorophenylboronic acid, Sigma-Aldrich®
:
Merck
3,5-Difluorophenylboronic acid, Sigma-Aldrich®
:
Merck
:
156545-07-2
:
:
C₆H₅BF₂O₂
:
MFCD01318138
:
157.91 g/mol
:
M-Clarity™ quality level = MQ100
:
GHS07