3,4-Difluorophenylboronic acid ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 168267-41-2 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: 300° Beilstein Reference: 7369788
Synonyms: 3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane
- CAS Number:168267-41-2
- Formula:C₆H₅BF₂O₂
- MDL Number:MFCD00807405
- Molecular Weight:157.91 g/mol
- Melting Pt:>300 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 168267-41-2 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: 300° Beilstein Reference: 7369788
Synonyms: 3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane
- CAS Number:168267-41-2
- Formula:C₆H₅BF₂O₂
- MDL Number:MFCD00807405
- Molecular Weight:157.91 g/mol
- Melting Pt:>300 °C
- Storage Temperature:Ambient



