Order Entry
Export
Orders LinkContactUsLinkComponent
2-Fluorophenylboronic acid

2-Fluorophenylboronic acid

Glass bottle for solids, 1 g
Catalog # MOLE18550854-1G
Supplier:   Molekula
2-Fluorophenylboronic acid
Catalog # MOLE18550854-1G
Supplier:  Molekula
CAS Number:  1993-03-9
2-Fluorophenylboronic acid

Glass bottle for solids, 1 g

CAS Number:  1993-03-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid
  • CAS Number:1993-03-9
  • Formula:C₆H₆BFO₂
  • MDL Number:MFCD00674013
  • Molecular Weight:139.92 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications