Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluorophenylboronic acid
2,6-Difluorophenylboronic acid, Sigma-Aldrich®
2,6-Difluorophenylboronic acid, Sigma-Aldrich®
:
Merck
2,6-Difluorophenylboronic acid, Sigma-Aldrich®
:
Merck
:
162101-25-9
:
:
C₆H₅BF₂O₂
:
MFCD00792436
:
M-Clarity™ quality level = MQ100
:
157.91 g/mol
:
GHS07