1,4-Difluorobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:540-36-3
:
Formula:C₆H₄F₂
MDL Number:MFCD00000344
:114.09 g/mol
Merck Index:13,03172
Boiling Pt:88 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:208-742-9