Potassium hydrogen phthalate 99.99% acidimetric standard
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:877-24-7
:
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070
:204.22 g/mol
Merck Index:13,07694
Melting Pt:295…300 °C (dec.)
Density:1,6362 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-889-4
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Assay:99.95 to 100.05 %
- pH:3.8 to 4 (5 % at 20°C)
- Sulfate (SO4):≤60 ppm
- Chlorine compounds:≤30 ppm (as Cl)
- Iron (Fe):≤5 ppm
- Sodium (Na):≤50 ppm
- Heavy metals (as Pb):≤5 ppm