3,5-Diaminobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:535-87-5
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007807
:152.15 g/mol
Merck Index:13,02997
Boiling Pt:546 °C (1013 hPa)
Melting Pt:239…242 °C
Density:1.45 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:208-621-0