
3,4-Diaminobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:619-05-6
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007726
:152.15 g/mol
Boiling Pt:354 °C (1013 hPa)
Melting Pt:211 °C
Density:1.544 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:210-577-2
:GHS07
Storage Temperature:Ambient