
4,5-Dimethoxyanthranilic acid, Sigma-Aldrich®
Catalog # SIAL252042-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:197.19 g/mol
Boiling Pt:359 °C (1013 hPa)
Melting Pt:155…160 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:227-095-3
:GHS07
CAS Number:5653-40-7
Formula:C₉H₁₁NO₄
MDL Number:MFCD00011671