4-Aminobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:150-13-0
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007894
:137.14 g/mol
Merck Index:13,00422
Boiling Pt:340 °C (1013 hPa)
Melting Pt:188 °C
Density:1.45 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:205-753-0