Benzoic acid
Plastic bottle for solids, 1
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
- CAS Number:65-85-0
- Formula:C₇H₆O₂
- MDL Number:MFCD00002398
- Molecular Weight:122.12 g/mol
- Merck Index:13,01092
- Boiling Pt:249 °C (1013 hPa)
- Melting Pt:121…123 °C
- Density:1.316 g/cm³ (20 °C)
- Flash Pt:121 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-618-2
- REACH:01-2119455536-33
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:1



