Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl phthalate

Diethyl phthalate

Glass bottle for solids, 100 mg
Catalog # PROOMM0006.28
Diethyl phthalate
Catalog # PROOMM0006.28
CAS Number:  84-66-2
Diethyl phthalate
Diethyl phthalate

Glass bottle for solids, 100 mg

CAS Number:  84-66-2
Price
615,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Phthalic acid diethyl ester
  • CAS Number:84-66-2
  • Formula:C₁₂H₁₄O₄
  • MDL Number:MFCD00009111
  • Molecular Weight:222.24 g/mol
  • Merck Index:12,07527
  • Boiling Pt:298 °C (1013 hPa)
  • Melting Pt:–3 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:156 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:201-550-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications