Propyl-4-hydroxybenzoate, Sigma-Aldrich®
SIAL54789-1KG-F
: Merck
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
:
:180.2 g/mol
:13,07958
:292 °C (1013 hPa)
:95 °C
:1.28 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:202-307-7
:GHS07
:94-13-3
:C₁₀H₁₂O₃
:MFCD00002354