Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-76-3
:
Formula:HOC₆H₄CO₂CH₃
MDL Number:MFCD00002352
:152.15 g/mol
Merck Index:13,06129
Boiling Pt:270…280 °C (1013 hPa)
Melting Pt:125…128 °C
Density:1.1208 g/cm³ (137 °C)
:M-Clarity™ quality level = MQ200
EINECS:202-785-7
:GHS07
Storage Temperature:Ambient