Order Entry
Export
ContactUsLinkComponent
Methyl-4-hydroxybenzoate 98.0-102.0% Ph. Eur.
Methyl-4-hydroxybenzoate 98.0-102.0% Ph. Eur.
Catalog # 25604.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Methyl-4-hydroxybenzoate 98.0-102.0% Ph. Eur.
Catalog # 25604.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98,0 - 102,0 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Melting point:
    125 - 128 °C
  • Identification (IR):
    Passes test
  • Appearance of solution:
    Passes test
  • Acidity:
    Passes test
  • 4-Hydroxybenzoic acid:
    ≤ 0,5 %
  • Unspecified impurities (for each):
    ≤ 0,5 %
  • Total impurities:
    ≤ 1,0 %
  • Disregard limit:
    ≤ 0,1 %
  • Sulphated ash:
    ≤ 0,1 %
  • Residual solvents:
    Passes test
  • As (Arsenic):
    ≤ 2 ppm
  • Cd (Cadmium):
    ≤ 0.5 ppm
  • Co (Cobalt):
    ≤ 0.5 ppm
  • Hg (Mercury):
    ≤ 2 ppm
  • Ni (Nickel):
    ≤ 0.5 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • V (Vanadium):
    ≤ 0.5 ppm
  • Conforms to Ph.Eur.:
    Passes test

Specification Test Results

About this item

 :
 :152.15 g/mol
Merck Index:13,06129
Boiling Pt:270…280 °C (1013 hPa)
Melting Pt:125…128 °C
Density:1.1208 g/cm³ (137 °C)
Storage Temperature:Ambient
EINECS:202-785-7
 :GHS07
CAS Number:99-76-3
Formula:HOC₆H₄CO₂CH₃
MDL Number:MFCD00002352