Order Entry
Export
ContactUsLinkComponent
Ethyl 4-hydroxybenzoate
Ethyl 4-hydroxybenzoate
Catalog # ROTH4746.1
Supplier:  Roth Carl
CAS Number:  
Ethyl 4-hydroxybenzoate
Catalog # ROTH4746.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
Merck Index:13,03869
Boiling Pt:297 °C (1013 hPa)
Melting Pt:116…118 °C
Density:1.078 g/cm³ (131 °C)
 :166.18 g/mol
Storage Temperature:Ambient
EINECS:204-399-4
CAS Number:120-47-8
Formula:C₉H₁₀O₃
MDL Number:MFCD00002353