Methyl anthranilate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:134-20-3
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007710
:151.16 g/mol
Merck Index:12,06099
Boiling Pt:237 °C (1013 hPa)
Melting Pt:24 °C
Density:1.16 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:205-132-4