Salicylamide ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: 2-Hydroxybenzamide
CAS Number:65-45-2
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007978
Boiling Pt:318
Melting Pt:137
Density:1.33 g/cm³ (20 °C)
Molecular Weight:137.14 g/mol
Merck Index:12,08480
EINECS:200-609-3
:GHS07
Specifications
- Environmentally Preferable: