
3,4-Dimethoxybenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3535-37-3
:
Formula:C₉H₉ClO₃
MDL Number:MFCD00000674
:200.62 g/mol
Boiling Pt:95…98 °C (1 mmHg)
Melting Pt:71 °C
EINECS:222-568-0
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05