Order Entry
Export
ContactUsLinkComponent
3,4-Dimethoxybenzoyl chloride 98%
Irritant
Corrosive
3,4-Dimethoxybenzoyl chloride 98%
Certificates
undefined
Irritant
Corrosive
3,4-Dimethoxybenzoyl chloride 98%

Specification Test Results

  • Appearance:
    White to yellow to grey Powder or crystals or flakes and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 74 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information