Order Entry
Export
ContactUsLinkComponent
3,5-Dinitrobenzoyl chloride 99%
Health
Corrosive
3,5-Dinitrobenzoyl chloride 99%
Certificates
undefined
Health
Corrosive
3,5-Dinitrobenzoyl chloride 99%
Certificates

Specification Test Results

  • Appearance:
    Yellow to beige Crystals or crystalline needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    67 to 71 °C
  • Titration RCOCl:
    ≥98.5 %

Specification Test Results

Related Information