Order Entry
Export
ContactUsLinkComponent
4-Nitrophthalonitrile 99%
Irritant
4-Nitrophthalonitrile 99%
Certificates
undefined
Irritant
4-Nitrophthalonitrile 99%

Specification Test Results

  • Appearance:
    Light yellow to beige to light green to light grey Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    140 to 144 °C
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information