3,4-Dicyanoaniline
Catalog # MOLE32596637
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:321 °C (1013 hPa)
Melting Pt:180…182 °C
UN:3439
ADR:6.1,III
:143.15 g/mol
EINECS:260-370-6
CAS Number:56765-79-8
Formula:C₈H₅N₃
MDL Number:MFCD00041741