3,4-Dihydroxybenzonitrile, Sigma-Aldrich®
Catalog # SIAL538396-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:311 °C (1013 hPa)
Melting Pt:160…163 °C
EINECS:241-367-9
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:135.12 g/mol
UN:3439
CAS Number:17345-61-8
Formula:C₇H₅NO₂
MDL Number:MFCD00016436