3,4-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-31-7
:
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00041255
:245.51 g/mol
Boiling Pt:158…160 °C (15 mmHg)
Melting Pt:24.2…24.8 °C
Density:1.572 g/cm³ (20 °C)
EINECS:202-656-5
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265