Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichlorobenzenesulphonyl chloride
2,6-Dichlorobenzenesulphonyl chloride
2,6-Dichlorobenzenesulphonyl chloride
2,6-Dichlorobenzenesulphonyl chloride
Supplier:
Thermo Scientific
Certificates
Expand image
2,6-Dichlorobenzenesulphonyl chloride
Supplier:
Thermo Scientific
About this item
CAS Number:
6579-54-0
:
Formula:
C₆H₃Cl₃O₂S
MDL Number:
MFCD00052311
Melting Pt:
54…56 °C
UN:
3261
ADR:
8,II
:
245.51 g/mol