Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichlorobenzenesulphonyl chloride
2,4-Dichlorobenzenesulphonyl chloride
2,4-Dichlorobenzenesulphonyl chloride
:
Thermo Scientific
2,4-Dichlorobenzenesulphonyl chloride
:
Thermo Scientific
:
16271-33-3
:
:
C₆H₃Cl₃O₂S
:
MFCD00052712
:
245.51 g/mol
:
56 °C
:
3261
:
8,II