2,3-Dichlorobenzenesulphonyl chloride
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:82417-45-6
:
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00051844
:245.51 g/mol
Boiling Pt:140…142 °C (2 mmHg)
Melting Pt:61…63 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II