Order Entry
Export
Orders LinkContactUsLinkComponent
4-Chlorothioanisole

4-Chlorothioanisole

Irritant
Supplier:   Thermo Scientific
4-Chlorothioanisole
Supplier:  Thermo Scientific
Certificates
4-Chlorothioanisole

3 Options Available Below

Product Details & Documents
Synonyms:  1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
  • CAS Number:123-09-1
  • Formula:C₇H₇ClS
  • MDL Number:MFCD00013643
  • Molecular Weight:158.65 g/mol
  • Boiling Pt:44 °C (0,08 mmHg)
  • Boiling Pt:44 °C (0.08 mmHg)
  • Melting Pt:27…28 °C
  • Density:1,2151 g/cm³ (19 °C)
  • EINECS:204-600-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g