Order Entry
Export
Orders LinkContactUsLinkComponent
Pentachlorophenol, Sigma-Aldrich®

Pentachlorophenol, Sigma-Aldrich®

Toxic
Health
Environment
Supplier:   Merck
Pentachlorophenol, Sigma-Aldrich®
Supplier:  Merck
Certificates
Pentachlorophenol, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:87-86-5
  • Formula:C₆HCl₅O
  • Molecular Weight:266.34 g/mol
  • Merck Index:12,07242
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:188 °C
  • Density:1.98 g/cm³ (20 °C)
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ200
  •  :Density: 1.98 g/cm³ (20 °C)
  • EINECS:201-778-6
  • UN:3155
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 38,40 €
    38,40 €
    1 * 5 g
       38,40 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 137,00 €
    137,00 €
    1 * 100 g
       137,00 €