
3,5-Dichlorophenol, Sigma-Aldrich®
Catalog # SIALD70600-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:163 g/mol
Boiling Pt:233 °C (1013 hPa)
Melting Pt:67.8 °C
Storage Temperature:Ambient
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:209-714-9
UN:2020
:GHS07
CAS Number:591-35-5
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002259