Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Dichlorophenol, Sigma-Aldrich®

Toxic
Corrosive
Environment
Supplier:   Merck
2,4-Dichlorophenol, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,4-Dichlorophenol, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene
  • CAS Number:120-83-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002169
  • Molecular Weight:163 g/mol
  • Merck Index:13,03098
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:40…41 °C
  • Density:1.4 g/cm³ (20 °C)
  • Flash Pt:114 °C
  • Storage Temperature:Ambient
  • UN:2928
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:204-429-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 31,50 €
    31,50 €
    1 * 5 g
       31,50 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 34,90 €
    34,90 €
    1 * 100 g
       34,90 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 82,70 €
    82,70 €
    1 * 250 g
       82,70 €