Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichlorophenol
2,6-Dichlorophenol, Sigma-Aldrich®
2,6-Dichlorophenol, Sigma-Aldrich®
:
Merck
2,6-Dichlorophenol, Sigma-Aldrich®
:
Merck
:
87-65-0
:
:
C₆H₄Cl₂O
:
MFCD00002176
:
163 g/mol
:
13,03099
:
∼219 °C (1013 hPa)
:
68 °C
:
1.653 g/cm³ (20 °C)
:
2020
:
6.1,III
:
M-Clarity™ quality level = MQ200
:
201-761-3
:
GHS07
:
Ambient