3,4-Dichlorophenol, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:95-77-2
- Formula:C₆H₄Cl₂O
- MDL Number:MFCD00002258
- Molecular Weight:163 g/mol
- Boiling Pt:247 °C (1013 hPa)
- Melting Pt:66…68 °C
- Density:1.6 g/cm³ (20 °C)
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- :Density: 1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-450-5
- UN:2020
2 Options Available Below
Product Details & Documents
- CAS Number:95-77-2
- Formula:C₆H₄Cl₂O
- MDL Number:MFCD00002258
- Molecular Weight:163 g/mol
- Boiling Pt:247 °C (1013 hPa)
- Melting Pt:66…68 °C
- Density:1.6 g/cm³ (20 °C)
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- :Density: 1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-450-5
- UN:2020



