
2,3-Dichlorophenol, Sigma-Aldrich®
Catalog # SIALD69807-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:163 g/mol
Boiling Pt:206 °C (13 hPa)
Melting Pt:57…58 °C
Density:1.4 g/cm³ (25 °C)
Flash Pt:115 °C
Storage Temperature:Ambient
EINECS:209-399-8
UN:2020
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:576-24-9
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002160