1-Chloro-3-nitrobenzene
Catalog # EHERC11453600
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:157.56 g/mol
Merck Index:13,02170
Boiling Pt:236 °C (1013 hPa)
Melting Pt:46 °C
Density:1.343 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-496-1
UN:1578
ADR:6.1,II
CAS Number:121-73-3
Formula:C₆H₄ClNO₂
MDL Number:MFCD00007202