
4-Amino-2,6-dichlorophenol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5930-28-9
:
Formula:C₆H₅Cl₂NO
MDL Number:MFCD00007875
:178.02 g/mol
Boiling Pt:304 °C (1013 hPa)
Melting Pt:164…168 °C
EINECS:227-671-4
:GHS07
Specification Test Results
- Appearance:Beige to light brown to light red Powder
- Infrared spectrum:Conforms
- Melting point:164 to 168 °C
- Solubility:(2.5 % in 96 % ethanol) Clear to slightly hazy
- Titration with TBAH:≥97.5 %