5-Chloro-2-nitroaniline, Sigma-Aldrich®
Catalog # SIAL269050-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:172.57 g/mol
Boiling Pt:323 °C (1013 hPa)
Melting Pt:123…126 °C
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:216-661-5
UN:2237
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:1635-61-6
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD00007776