Order Entry
Export
ContactUsLinkComponent
3,5-Dichlorophenyl isocyanate, Sigma-Aldrich®
3,5-Dichlorophenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL389056-5G
Supplier:  Merck
CAS Number:  
3,5-Dichlorophenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL389056-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :188.01 g/mol
Boiling Pt:264 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:252-276-9
UN:2250
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
CAS Number:34893-92-0
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00013859