2,4-Dichlorophenyl isocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:2612-57-9
:
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00002001
:188.01 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:59…61 °C
Storage Temperature:Refrigerator
EINECS:220-040-4
UN:2250
ADR:6.1,II