3,4-Dichlorophenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL245607-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Boiling Pt:234 °C (1013 hPa)
Melting Pt:38…41 °C
EINECS:203-026-2
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:188.01 g/mol
UN:2250
CAS Number:102-36-3
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00002017