1,2-Dichloro-4-iodobenzene, Sigma-Aldrich®
Catalog # SIAL541753-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:272.9 g/mol
:M-Clarity™ quality level = MQ100
EINECS:243-874-0
CAS Number:20555-91-3
Formula:C₆H₃Cl₂I
MDL Number:MFCD00019014