2-Chloro-3,4-dimethoxybenzoic acid, Sigma-Aldrich®
Catalog # SIAL638730-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.62 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:52009-53-7
Formula:C₉H₉ClO₄
MDL Number:MFCD00084944