5-Chloro-2-methoxybenzoic acid, Sigma-Aldrich®
Catalog # SIAL292494-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:186.59 g/mol
:M-Clarity™ quality level = MQ100
EINECS:222-343-7
CAS Number:3438-16-2
Formula:C₈H₇ClO₃
MDL Number:MFCD00017546