Order Entry
Export
ContactUsLinkComponent
 
5-Chloro-2-methoxybenzoic acid
Catalog # PROOMM0008.03
CAS Number:  
5-Chloro-2-methoxybenzoic acid
Catalog # PROOMM0008.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :186.59 g/mol
EINECS:222-343-7
CAS Number:3438-16-2
Formula:C₈H₇ClO₃
MDL Number:MFCD00017546